Novus Biologicals
Manufacturer Code:NBP172140
Catalog # NBP172140
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 19 amino acid synthetic peptide near the amino of terminus human PPAPDC3. The immunogen is located within the first 50 amino acids of PPAPDC3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C9orf67 chromosome 9 open reading frame 67 EC 3.1.3 EC 3.1.3.- FLJ14662 KIAA0515 MGC12921 NET39 phosphatidic acid phosphatase type 2 domain containing 3 Phosphatidic acid phosphatase type 2 domain-containing protein 3 probable lipid phosphate phosphatase PPAPDC3; Inactive phospholipid phosphatase 7; nuclear envelope transmembrane protein NET39; phosphatidic acid phosphatase type 2 domain containing 3; Phosphatidic acid phosphatase type 2 domain-containing protein 3; probable lipid phosphate phosphatase PPAPDC3
Gene Aliases: C9orf67; NET39; PLPP7; PPAPDC3
UniProt ID: (Human) Q8NBV4
Entrez Gene ID: (Human) 84814
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.