Proteintech
Manufacturer Code:206351AP150UL
Catalog # 50-173-3004
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PPAPDC3 Fusion Protein Ag14676 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C9orf67 KIAA0515 NET39 PPAPDC3; Inactive phospholipid phosphatase 7; Nuclear envelope transmembrane protein 39; nuclear envelope transmembrane protein NET39; phosphatidic acid phosphatase type 2 domain containing 3; Phosphatidic acid phosphatase type 2 domain-containing protein 3; probable lipid phosphate phosphatase PPAPDC3
Gene Aliases: C9orf67; D830019K17Rik; NET39; PLPP7; PPAPDC3; RGD1305821
UniProt ID: (Human) Q8NBV4, (Rat) Q5FVJ3, (Mouse) Q91WB2
Entrez Gene ID: (Human) 84814, (Rat) 296635, (Mouse) 227721
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.