Invitrogen
Manufacturer Code:PA520865
Catalog # PIPA520865
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 13 amino acid peptide from near the carboxy terminus human PPAPDC2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: phosphatidic acid phosphatase type 2 domain containing 2; Phosphatidic acid phosphatase type 2 domain-containing protein 2; Phospholipid phosphatase 6; polyisoprenoid diphosphate phosphatase type 1; PPAP2 domain-containing protein 2; Presqualene diphosphate phosphatase; RP11-6J24.2 PDP1 PSDP bA6J24.6 PPAP2 domain-containing protein 2 phosphatidic acid phosphatase type 2 domain-containing protein 2 polyisoprenoid diphosphate phosphatase type 1 presqualene diphosphate phosphatase
Gene Aliases: 4932443D16Rik; bA6J24.6; PDP1; PLPP6; PPAPDC2; PSDP
UniProt ID: (Human) Q8IY26, (Mouse) Q9D4F2, (Rat) Q66H88
Entrez Gene ID: (Human) 403313, (Mouse) 74411, (Rat) 619549
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.