Abnova
Manufacturer Code:PAB21010
Catalog # 89-122-506
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human PPAPDC2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant PPAPDC2.
For Research Use Only
Protein Aliases: bA6J24.6; FLJ46512; FLJ90191; MGC15483; phosphatidic acid phosphatase type 2 domain containing 2; Phosphatidic acid phosphatase type 2 domain-containing protein 2; Phospholipid phosphatase 6; polyisoprenoid diphosphate phosphatase type 1; PPAP2 domain-containing protein 2; Presqualene diphosphate phosphatase; PSDP
Gene Aliases: bA6J24.6; PDP1; PLPP6; PPAPDC2; PSDP
UniProt ID: (Human) Q8IY26
Entrez Gene ID: (Human) 403313
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.