Abnova
Manufacturer Code:H00008613A01
Catalog # 89-105-899
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 1 a.a. &bsim 311 a.a) full-length recombinant protein with GST tag.,PPAP2B (AAH09196 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length recombinant PPAP2B.
For Research Use Only
Protein Aliases: Dri42; Lipid phosphate phosphohydrolase 3; LPP3; MGC15306; PAP-2b; PAP2 beta; PAP2-b; PAP2-beta; Phosphatidate phosphohydrolase type 2b; Phosphatidic acid phosphatase 2b; phosphatidic acid phosphatase type 2B; Phospholipid phosphatase 3; type-2 phosphatidic acid phosphatase-beta; vascular endothelial growth factor and type I collagen inducible; Vascular endothelial growth factor and type I collagen-inducible protein; VCIP
Gene Aliases: Dri42; LPP3; PAP2B; PLPP3; PPAP2B; VCIP
UniProt ID: (Human) O14495
Entrez Gene ID: (Human) 8613
Molecular Function: hydrolase phosphatase pyrophosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.