Novus Biologicals
Manufacturer Code:NBP245378
Catalog # NBP245378
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1H4 |
Immunogen | Full length human recombinant protein of human PPAP2A(NP_003702) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.3.4 lipid phosphate phosphohydrolase 1 lipid phosphate phosphohydrolase 1a LLP1a LPP1PAP2a2 PAP2 PAP2a PAP2-alpha PAP-2aPAP2alpha2 PAPalpha1 Phosphatidate phosphohydrolase type 2a Phosphatidic acid phosphatase 2a phosphatidic acid phosphatase type 2A phosphatidic acid phosphohydrolase type 2a type 2 phosphatidic acid phosphohydrolase type-2 phosphatidic acid phosphatase alpha; Lipid phosphate phosphohydrolase 1; lipid phosphate phosphohydrolase 1a; PAP-2a; PAP2-alpha; Phosphatidate phosphohydrolase type 2a; Phosphatidic acid phosphatase 2a; phosphatidic acid phosphatase type 2A; phosphatidic acid phosphohydrolase type 2a; Phospholipid phosphatase 1; type-2 phosphatidic acid phosphatase alpha
Gene Aliases: LLP1a; LPP1; PAP-2a; PAP2; PLPP1; PPAP2A
UniProt ID: (Human) O14494
Entrez Gene ID: (Human) 8611
Molecular Function:
hydrolase
phosphatase
pyrophosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.