Novus Biologicals
Manufacturer Code:NBP28985750UL
Catalog # NB021181
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1 |
Immunogen | Recombinant Human PP2C gamma/PPM1G protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.3.16 MGC1675 MGC2870 PP2C gamma PP2CG PP2C-gamma PP2CGAMMA PPM1C PPP2CG Protein phosphatase 1C protein phosphatase 1G protein phosphatase 1G (formerly 2C) magnesium-dependent gamma isoform protein phosphatase 2 catalytic subunit gamma isoform protein phosphatase 2C gamma isoform Protein phosphatase 2C isoform gamma Protein phosphatase magnesium-dependent 1 gamma protein phosphatase Mg2+/Mn2+ dependent 1G; PP2C, gamma; PP2C-gamma; Protein phosphatase 1C; Protein phosphatase 1G; protein phosphatase 1G (formerly 2C), magnesium-dependent, gamma isoform; protein phosphatase 2, catalytic subunit, gamma isoform; protein phosphatase 2C gamma isoform; Protein phosphatase 2C isoform gamma; Protein phosphatase magnesium-dependent 1 gamma; protein phosphatase, Mg2+/Mn2+ dependent, 1G
Gene Aliases: PP2CG; PP2CGAMMA; PPM1C; PPM1G; PPP2CG
UniProt ID: (Human) O15355
Entrez Gene ID: (Human) 5496
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.