Novus Biologicals
Manufacturer Code:NBP298253AF532
Catalog # NB053005
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant PP2C alpha/PPM1A (NP_032936.1 Met 1-Trp 382). |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.3.16 FLJ42306 MGC9201 phosphatase 2C alpha PP2CA PP2Calpha PP2C-ALPHA PPPM1A protein phosphatase 1A protein phosphatase 1A (formerly 2C) magnesium-dependent alpha isoform protein phosphatase 2C alpha isoform Protein phosphatase 2C isoform alpha Protein phosphatase IA protein phosphatase Mg2+/Mn2+ dependent 1A; PP2C-alpha; Protein phosphatase 1A; protein phosphatase 1A (formerly 2C), magnesium-dependent, alpha isoform; Protein phosphatase 2C isoform alpha; Protein phosphatase IA; protein phosphatase, Mg2+/Mn2+ dependent, 1A
Gene Aliases: PP2C-ALPHA; PP2CA; PP2Calpha; PPM1A; PPPM1A
UniProt ID: (Human) P35813
Entrez Gene ID: (Human) 5494
Molecular Function: enzyme modulator hydrolase kinase inhibitor kinase modulator phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.