Invitrogen
Manufacturer Code:MA538160
Catalog # PIMA538160
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1889 |
Immunogen | A synthesized peptide derived from human PP2A alpha + beta |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: AI115466 D8Ertd766e fb17h07 fc05c02 fk24a01 hypothetical protein LOC550585 PP2A Pp2a1 Pp2a2 PP2A-alpha PP2Abeta PP2A-beta PP2Ac PP2CA PP2Calpha PP2CB PPP2CA PPP2CB protein phosphatase 2 (formerly 2A) catalytic subunit alpha isoform protein phosphatase 2 (formerly 2A) catalytic subunit beta isoform protein phosphatase 2 catalytic subunit alpha protein phosphatase 2 catalytic subunit beta protein phosphatase 2 catalytic subunit alpha isoform protein phosphatase 2 catalytic subunit alpha isozyme protein phosphatase 2 catalytic subunit beta isoform protein phosphatase 2 catalytic subunit beta isozyme protein phosphatase 2catalytic subunit alpha isozyme protein phosphatase 2A alpha subunit protein phosphatase 2A beta subunit protein phosphatase 2A catalytic alpha subunit protein phosphatase 2A catalytic subunit alpha isoform protein phosphatase 2A catalytic subunit beta isoform protein phosphatase 2A catalytic subunit beta isoform protein phosphatase 2a catalytic subunit alpha isoform protein phosphatase 2A catalytic subunit beta isoform protein phosphatase type 2A catalytic subunit protein phosphatase-2A-alpha protein phosphatase-2A-beta R75353 Replication protein C RP C RP-C serine/threonine protein phosphatase 2A catalytic subunit alpha isoform serine/threonine protein phosphatase 2A catalytic subunit beta isoform serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform Serine/threonine-protein phosphatase 2A catalytic subunit beta isoform testicular tissue protein Li 146 wu:fb17h07 wu:fc05c02 wu:fk24a01 zgc:110641 zgc:55808 zgc:77172; PP2A-alpha; PP2A-beta; protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform; protein phosphatase 2 (formerly 2A), catalytic subunit, beta isoform; protein phosphatase 2, catalytic subunit, alpha isoform; protein phosphatase 2, catalytic subunit, alpha isozyme; protein phosphatase 2, catalytic subunit, beta isoform; protein phosphatase 2, catalytic subunit, beta isozyme; protein phosphatase 2A catalytic alpha subunit; protein phosphatase 2A catalytic subunit, beta isoform; protein phosphatase 2a, catalytic subunit, alpha isoform; protein phosphatase 2a, catalytic subunit, beta isoform; protein phosphatase type 2A catalytic subunit; protein phosphatase-2A-alpha; protein phosphatase-2A-beta; Replication protein C; RP-C; serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform; serine/threonine protein phosphatase 2A, catalytic subunit, beta isoform; Serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform; Serine/threonine-protein phosphatase 2A catalytic subunit beta isoform; testicular tissue protein Li 146
Gene Aliases: AI115466; D8Ertd766e; PP2A; Pp2a1; Pp2a2; PP2Abeta; PP2Ac; PP2CA; PP2Calpha; PP2CB; PPP2CA; PPP2CB; R75353; RP-C
UniProt ID: (Human) P67775, (Human) P62714, (Mouse) P63330, (Rat) P63331, (Rat) P62716, (Mouse) P62715
Entrez Gene ID: (Human) 5515, (Human) 5516, (Mouse) 19052, (Rat) 24672, (Rat) 24673, (Mouse) 19053
Molecular Function:
calcium-binding protein
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.