Invitrogen
Manufacturer Code:MA535878
Catalog # PIMA535878
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1589 |
Immunogen | A phospho specific peptide corresponding to residues surrounding Y307 of human PP2A alpha |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: B55-GAMMA BOS_8121 IMYPNO IMYPNO1 Methyl-PPP2CA MGC33570 PP2A PP2A alpha PP2A C terminal PP2A L9 PP2A Leucine 9 Pp2a1 PP2A-alpha PP2Ac PP2CA PP2Calpha Ppp2ca PR52 PR55G R75353 RCJMB04_3c21 Replication protein C RP-C Serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform; PP2A-alpha; protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform; protein phosphatase 2, catalytic subunit, alpha isozyme; protein phosphatase 2a, catalytic subunit, alpha isoform; Replication protein C; RP-C; serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform; Serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform
Gene Aliases: PP2A; PP2Ac; PP2CA; PP2Calpha; PPP2CA; R75353; RP-C
UniProt ID: (Human) P67775, (Mouse) P63330
Entrez Gene ID: (Human) 5515, (Mouse) 19052
Molecular Function: calcium-binding protein hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.