Novus Biologicals
Manufacturer Code:NBP229750
Catalog # NBP229750
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4G7 |
Immunogen | Peptide corresponding to amino acids 6-24 of human Protein Phosphatase 2A A subunit. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: EC 3.1.3.16 PP2A-alpha PP2Ac PP2CA PP2Calpha protein phosphatase 2 (formerly 2A) catalytic subunit alpha isoform protein phosphatase 2 catalytic subunit alpha isozyme protein phosphatase 2A catalytic subunit alpha isoform Replication protein C RP-C serine/threonine protein phosphatase 2A catalytic subunit alpha isoform serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform; PP2A-alpha; protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform; protein phosphatase 2, catalytic subunit, alpha isozyme; Replication protein C; RP-C; serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform; Serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform
Gene Aliases: PP2Ac; PP2CA; PP2Calpha; PPP2CA; RP-C
UniProt ID: (Human) P67775
Entrez Gene ID: (Human) 5515
Molecular Function: calcium-binding protein hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.