Invitrogen
Manufacturer Code:PA112393
Catalog # 01-674-126
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: B55-GAMMA IMYPNO IMYPNO1 MGC33570 PR52 PR55G; PP2A-alpha; protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform; protein phosphatase 2, catalytic subunit, alpha isoform; protein phosphatase 2, catalytic subunit, alpha isozyme; protein phosphatase 2a, catalytic subunit, alpha isoform; protein phosphatase type 2A catalytic subunit; protein phosphatase-2A-alpha; Replication protein C; RP-C; serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform; Serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform; unnamed protein product
Gene Aliases: PP2A; Pp2a1; PP2Ac; PP2CA; PP2Calpha; PPP2CA; R75353; RP-C
UniProt ID: (Human) P67775, (Bovine) P67774, (Mouse) P63330, (Rat) P63331
Entrez Gene ID: (Human) 5515, (Bovine) 282320, (Mouse) 19052, (Rat) 24672
Molecular Function:
calcium-binding protein
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.