Invitrogen
Manufacturer Code:PA578116
Catalog # PIPA578116
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human POU4F3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Brain-3C; brain-specific homeobox POU domain protein 3C; Brain-specific homeobox/POU domain protein 3C; Brn-3.1; brn-3C; DFNA15 BRN3C POU class 4 homeobox 3; POU domain, class 4, transcription factor 3
Gene Aliases: Brn-3.1; Brn-3c; Brn3.1; BRN3C; ddl; DFNA15; dreidel; POU4F3
UniProt ID: (Human) Q15319, (Mouse) Q63955
Entrez Gene ID: (Human) 5459, (Mouse) 18998
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.