Invitrogen
Manufacturer Code:PA540644
Catalog # PIPA540644
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human POU4F3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: brain POU domain gene 3.1; Brain-3.1; Brain-3C; brain-specific homeobox POU domain protein 3C; Brain-specific homeobox/POU domain protein 3.1; Brain-specific homeobox/POU domain protein 3C; Brn-3.1; brn-3C; POU class 4 homeobox 3 BRN3C DFNA15; POU domain, class 4, transcription factor 3; zfbrn-3.1
Gene Aliases: BRN-3.1; Brn-3c; Brn3.1; BRN3C; ddl; DFNA15; dreidel; POU4F3
UniProt ID: (Human) Q15319, (Rat) D3ZTL1, (Mouse) Q63955, (Zebrafish) Q90435
Entrez Gene ID: (Bovine) 538520, (Dog) 487200, (Horse) 100071960, (Human) 5459, (Rabbit) 100357560, (Rat) 364855, (Mouse) 18998, (Guinea pig) 100713776, (Zebrafish) 30534
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.