Novus Biologicals
Manufacturer Code:NBP188349
Catalog # NBP188349
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:HQGLEGDLLEHISPTLSVSGLGAPEHSVMPAQIHPHHLGAMGHLHQAMGMSHPHTVAPHSAMPACLSDVESDPR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Brain-3C; brain-3C Brain-specific homeobox/POU domain protein 3C BRN3.1 Brn-3C BRN3Cbrn-3C DFNA15 MGC138412 POU class 4 homeobox 3 POU domain class 4 transcription factor 3 POU domain class 4 transcription factor 3; Brain-specific homeobox/POU domain protein 3C; brn-3C; POU domain, class 4, transcription factor 3
Gene Aliases: BRN3C; DFNA15; POU4F3
UniProt ID: (Human) Q15319
Entrez Gene ID: (Human) 5459
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.