Invitrogen
Manufacturer Code:PA595576
Catalog # PIPA595576
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human BRN3A recombit protein (Position: R311-Y419). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Brain-3A; Brain-specific homeobox/POU domain protein 3A; Brn-3.0; brn-3A; Homeobox/POU domain protein RDC-1; Oct-T1; POU domain class 4 transcription factor 1; POU domain, class 4, transcription factor 1
Gene Aliases: Brn-3; Brn-3.0; brn-3A; Brn3; Brn3.0; BRN3A; E130119J07Rik; Oct-T1; POU4F1; RDC-1; RDC1
UniProt ID: (Human) Q01851, (Mouse) P17208, (Rat) P20266
Entrez Gene ID: (Human) 5457, (Mouse) 18996, (Rat) 114503
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.