Novus Biologicals
Manufacturer Code:NBP183966
Catalog # NBP183966
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:LESMHTDIKMSGDVADSTDARSTLSQVEPGNDRNGLDFNRQIKTEDLSDSLQQTLSHRPCHLSQGPAMMSGNQMSGLNASPCQDMASLHPLQQLVLVPGHLQSVSQFLLSQTQP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Epoc-1 MGC126698 oct-11 OCT11FLJ40063 Octamer-binding protein 11 Octamer-binding transcription factor 11 OTF11 OTF-11 PLA1 PLA-1 POU class 2 homeobox 3 POU domain class 2 transcription factor 3 POU domain class 2 transcription factor 3 POU transcription factor Skn-1a Transcription factor PLA-1 Transcription factor Skn-1; Oct-11; Octamer-binding protein 11; Octamer-binding transcription factor 11; OTF-11; POU domain transcription factor OCT11a; POU domain, class 2, transcription factor 3; Transcription factor PLA-1; Transcription factor Skn-1
Gene Aliases: Epoc-1; OCT-11; OCT11; OTF-11; OTF11; PLA-1; PLA1; POU2F3; Skn-1a
UniProt ID: (Human) Q9UKI9
Entrez Gene ID: (Human) 25833
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.