R&D Systems
Manufacturer Code:MAB5299SP
Catalog # MAB5299SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 688923 |
Immunogen | <em>E. coli</em>-derived recombinant human POT1 Met159-Pro316 Accession # Q9NUX5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DKFZp586D211 hPot1 POT1 protection of telomeres 1 homolog POT1-like telomere end-binding protein protection of telomeres 1 homolog (S. pombe) protection of telomeres protein 1; hPot1; POT1-like telomere end-binding protein; protection of telomeres 1 homolog; Protection of telomeres protein 1
Gene Aliases: CMM10; GLM9; HPOT1; POT1
UniProt ID: (Human) Q9NUX5
Entrez Gene ID: (Human) 25913
Molecular Function:
DNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.