Invitrogen
Manufacturer Code:PA575366
Catalog # PIPA575366
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 269-315 of Human POT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hPot1; mPot1; POT1-like telomere end-binding protein; protection of telomeres 1 homolog; protection of telomeres 1A; Protection of telomeres protein 1; Protection of telomeres protein 1 hPot1 POT1-like telomere end-binding protein POT1
Gene Aliases: 1500031H18Rik; AI851169; CMM10; GLM9; HPOT1; POT1; Pot1a
UniProt ID: (Human) Q9NUX5, (Mouse) Q91WC1
Entrez Gene ID: (Human) 25913, (Rat) 500054, (Mouse) 101185
Molecular Function:
DNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.