Proteintech
Manufacturer Code:146491AP150UL
Catalog # 50-172-9615
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | POSH Fusion Protein Ag6243 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase SH3RF1; KIAA1494 Plenty of SH3s POSH Protein POSH RING finger protein 142 RNF142 SH3 multiple domains protein 2 SH3MD2 SH3RF1; plenty of SH3 domains; Plenty of SH3s; Protein POSH; putative E3 ubiquitin-protein ligase SH3RF1; putative scaffolding protein POSH; RING finger protein 142; RING-type E3 ubiquitin transferase SH3RF1; SH3 domain-containing RING finger protein 1; SH3 multiple domains 2; SH3 multiple domains protein 2
Gene Aliases: 2200003J05Rik; KIAA1494; POSH; POSH1; R75531; RNF142; SH3MD2; SH3RF1
UniProt ID: (Human) Q7Z6J0, (Rat) Q71F54, (Mouse) Q69ZI1
Entrez Gene ID: (Human) 57630, (Rat) 306417, (Mouse) 59009
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.