Invitrogen
Manufacturer Code:PA543423
Catalog # PIPA543423
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human PORCN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mPORC; porcupine homolog; Porcupine homolog (Drosophila) DHOF FODH MG61 MGC29687 PORC PPN por; probable protein-cysteine N-palmitoyltransferase porcupine; Protein MG61; protein-cysteine N-palmitoyltransferase porcupine; Protein-serine O-palmitoleoyltransferase porcupine
Gene Aliases: 2410004O13Rik; AW045557; DHOF; DXHXS7465e; FODH; MG61; mMg61; Mporc; PORC; PORCN; porcupine; PPN; RGD1564947
UniProt ID: (Human) Q9H237, (Mouse) Q9JJJ7
Entrez Gene ID: (Dog) 480903, (Rabbit) 100342483, (Human) 64840, (Bovine) 540373, (Horse) 100051598, (Mouse) 53627, (Rat) 317368, (Guinea pig) 100729593
Molecular Function: acetyltransferase membrane traffic protein membrane trafficking regulatory protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.