Novus Biologicals
Manufacturer Code:NBP275626
Catalog # NBP275626
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JB54-31 |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: CPR; CPR CYPORFLJ26468 DKFZp686G04235 EC 1.6.2.4 NADPH--cytochrome P450 reductase NADPH-dependent cytochrome P450 reductase P450 (cytochrome) oxidoreductase P450R; NADPH--cytochrome P450 reductase; NADPH-dependent cytochrome P450 reductase; P450 (cytochrome) oxidoreductase
Gene Aliases: CPR; CYPOR; P450R; POR
UniProt ID: (Human) P16435
Entrez Gene ID: (Human) 5447
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.