Invitrogen
Manufacturer Code:MA524443
Catalog # PIMA524443
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3F12 |
Immunogen | POP7 (NP&lowbar005828.1 1 a.a. &bsim 80 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C,Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: hPOP7; POP7 (processing of precursor, S. cerevisiae) homolog; POP7 RPP20; processing of precursor 7, ribonuclease P subunit; processing of precursor 7, ribonuclease P/MRP subunit; Ribonuclease P protein subunit p20; Ribonucleases P/MRP protein subunit POP7 homolog; RNaseP protein p20
Gene Aliases: 0610037N12Rik; POP7; RPP2; RPP20
UniProt ID: (Human) O75817
Entrez Gene ID: (Human) 10248
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.