Invitrogen
Manufacturer Code:PA570239
Catalog # PIPA570239
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human POP5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hPop5; processing of precursor 5, ribonuclease P/MRP family; processing of precursor 5, ribonuclease P/MRP subunit; ribonuclease P protein 3; Ribonuclease P/MRP protein subunit POP5; Ribonuclease P/MRP protein subunit POP5-like protein
Gene Aliases: 1500019J17Rik; 2700077E03Rik; AD-008; AW049989; fc69d03; hPop5; HSPC004; POP5; Rnasep3; RPP2; RPP20; wu:fc69d03; wu:fy24c05; x0003; zgc:110616
UniProt ID: (Human) Q969H6, (Bovine) Q1JQ92, (Mouse) Q9DB28, (Zebrafish) Q5BJI6
Entrez Gene ID: (Human) 51367, (Dog) 477515, (Horse) 100050189, (Rabbit) 100351855, (Bovine) 507410, (Rat) 117241, (Guinea pig) 100729028, (Mouse) 117109, (Zebrafish) 541421
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.