Novus Biologicals
Manufacturer Code:NBP298016JF646
Catalog # NB051034
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human POP4 fragment. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.26.5 processing of precursor 4 ribonuclease P/MRP subunit (S. cerevisiae) ribonuclease P protein subunit p29 RPP29hPOP4; hPOP4; Ribonuclease P protein subunit p29
Gene Aliases: POP4; RPP29
UniProt ID: (Human) O95707
Entrez Gene ID: (Human) 10775
Molecular Function:
RNA binding protein
endoribonuclease
esterase
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.