Novus Biologicals
Manufacturer Code:NBP126605
Catalog # NBP126605
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 75 and 125 of human processing of precursor 1 using the numbering given in entry NP_055844.1 (GeneID 10940). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.26.5 hPOP1 KIAA0061 MGC17365 processing of precursor 1 ribonuclease P/MRP subunit (S. cerevisiae) processing of precursors 1 ribonucleases P/MRP protein subunit POP1; hPOP1; processing of precursor 1, ribonuclease P/MRP subunit; processing of precursors 1; Ribonucleases P/MRP protein subunit POP1
Gene Aliases: KIAA0061; POP1
UniProt ID: (Human) Q99575
Entrez Gene ID: (Human) 10940
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.