Novus Biologicals
Manufacturer Code:NBP293628100UL
Catalog # NB029120
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human POP1 (NP_055844.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.26.5 hPOP1 KIAA0061 MGC17365 processing of precursor 1 ribonuclease P/MRP subunit (S. cerevisiae) processing of precursors 1 ribonucleases P/MRP protein subunit POP1; hPOP1; processing of precursor 1, ribonuclease P/MRP subunit; processing of precursors 1; Ribonucleases P/MRP protein subunit POP1
Gene Aliases: KIAA0061; POP1
UniProt ID: (Human) Q99575
Entrez Gene ID: (Human) 10940
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.