Proteintech
Manufacturer Code:120291AP150UL
Catalog # 50-172-7656
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | POP1 Fusion Protein Ag2660 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hPOP1; hPOP1 KIAA0061 POP1; processing of precursor 1, ribonuclease P/MRP subunit; processing of precursors 1; ribonucleases P/MRP protein subunit homolog; Ribonucleases P/MRP protein subunit POP1
Gene Aliases: 4932434G09Rik; KIAA0061; POP1
UniProt ID: (Human) Q99575
Entrez Gene ID: (Human) 10940, (Mouse) 67724, (Rat) 315045
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.