Invitrogen
Manufacturer Code:MA517219
Catalog # PIMA517219
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5G11 |
Immunogen | Recombit human protein purified from E.coli(His-PON3) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: arylesterase 3; paraoxanase-3; Serum paraoxonase/lactonase 3
Gene Aliases: PON3
UniProt ID: (Human) Q15166
Entrez Gene ID: (Human) 5446
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.