Invitrogen
Manufacturer Code:PA542068
Catalog # PIPA542068
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human PON3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: arylesterase 3; lactonase/esterase; paraoxanase-3; Paraoxonase 3; Serum paraoxonase/lactonase 3
Gene Aliases: 2810004E20; AI786302; PON3
UniProt ID: (Rabbit) Q9BGN0, (Human) Q15166, (Rat) Q68FP2, (Mouse) Q62087
Entrez Gene ID: (Rabbit) 100008754, (Human) 5446, (Dog) 475235, (Bovine) 510953, (Horse) 100051959, (Pig) 733674, (Rat) 312086, (Mouse) 269823
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.