R&D Systems
Manufacturer Code:AF4345
Catalog # AF4345
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human PON3 Ala30-Leu354 Accession # Q15166 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: arylesterase 3; EC 3.1.1.2 EC 3.1.1.81 EC 3.1.8.1 paraoxanase-3 paraoxonase 3 serum paraoxonase/lactonase 3; paraoxanase-3; Serum paraoxonase/lactonase 3
Gene Aliases: PON3
UniProt ID: (Human) Q15166
Entrez Gene ID: (Human) 5446
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.