Invitrogen
Manufacturer Code:MA524800
Catalog # PIMA524800
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4G8D3 |
Immunogen | Partial recombit human PON1 (between residues 1-200) expressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A-esterase 1; A-esterase 1 Aromatic esterase 1 arylesterase B-type EC 3.1.1.2 EC 3.1.1.81 EC 3.1.8.1 ESAesterase A K-45 MVCD5 paraoxonase 1 PON 1 PONparaoxonase B-type serum aryldiakylphosphatase Serum aryldialkylphosphatase 1 serum paraoxonase/arylesterase 1; Aromatic esterase 1; arylesterase 1; arylesterase B-type; esterase A; K-45; paraoxonase B-type; PON 1; serum aryldiakylphosphatase; Serum aryldialkylphosphatase 1; Serum paraoxonase/arylesterase 1
Gene Aliases: ESA; MVCD5; PON; PON1
UniProt ID: (Human) P27169
Entrez Gene ID: (Human) 5444
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.