Invitrogen
Manufacturer Code:PA579845
Catalog # PIPA579845
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived mouse PON1 recombit protein (Position: A30-D274). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A-esterase 1; Aromatic esterase 1; PON 1; Serum aryldialkylphosphatase 1; Serum paraoxonase/arylesterase 1; Serum paraoxonase/arylesterase 1 PON 1
Gene Aliases: Pon; Pon1
UniProt ID: (Mouse) P52430
Entrez Gene ID: (Mouse) 18979
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.