Novus Biologicals
Manufacturer Code:NBP219892
Catalog # NBP219892
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human POMZP3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC8359 POM (POM121 rat homolog) and ZP3 fusion POM121 and ZP3 fusion POM121 and ZP3 fusion protein POM121/ZP3 fusion protein rat) and ZP3 fusion; POM (POM121 homolog, rat) and ZP3 fusion; POM (POM121 rat homolog) and ZP3 fusion; POM-ZP3; POM-ZP3 fusion protein; POM121 and ZP3 fusion protein; POM121/ZP3 fusion protein
Gene Aliases: POM-ZP3; POM121; POMZP3
UniProt ID: (Human) F6STJ3
Entrez Gene ID: (Human) 22932
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.