Invitrogen
Manufacturer Code:PA551987
Catalog # PIPA551987
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human POMT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dolichyl-phosphate-mannose--protein mannosyltransferase; Dolichyl-phosphate-mannose--protein mannosyltransferase 2; Protein O-mannosyl-transferase 2; protein-O-mannosyltransferase 2; Protein-O-mannosyltransferase 2 DKFZp686G10254 FLJ22309 MDDGA2 MDDGB2 MDDGC2
Gene Aliases: A830009D15Rik; AW046274; LGMD2N; MDDGA2; MDDGB2; MDDGC2; POMT2
UniProt ID: (Human) Q9UKY4, (Mouse) Q8BGQ4
Entrez Gene ID: (Human) 29954, (Rat) 688673, (Mouse) 217734
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.