Invitrogen
Manufacturer Code:PA5113831
Catalog # PIPA5113831
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human POMT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A830009D15Rik AW046274 DKFZp686G10254 Dolichyl-phosphate-mannose--protein mannosyltransferase Dolichyl-phosphate-mannose--protein mannosyltransferase 2 FLJ22309 LGMD2N MDDGA2 MDDGB2 MDDGC2 Pomt2 protein O-mannosyltransferase 2 Protein O-mannosyl-transferase 2 protein-O-mannosyltransferase 2; Dolichyl-phosphate-mannose--protein mannosyltransferase; Dolichyl-phosphate-mannose--protein mannosyltransferase 2; Protein O-mannosyl-transferase 2; protein-O-mannosyltransferase 2
Gene Aliases: LGMD2N; MDDGA2; MDDGB2; MDDGC2; POMT2
UniProt ID: (Human) Q9UKY4
Entrez Gene ID: (Human) 29954
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.