Novus Biologicals
Manufacturer Code:NBP16006820UL
Catalog # NBP16006820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to POMT2(protein-O-mannosyltransferase 2) The peptide sequence was selected from the middle region of POMT2. Peptide sequence AIGYLHSHRHLYPEGIGARQQQVTTYLHKDYNNLWIIKKHNTNSDPLDPS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686G10254 Dolichyl-phosphate-mannose--protein mannosyltransferase 2 EC 2.4.1.109 FLJ22309 MDDGA2 MDDGB2 MDDGC2 protein O-mannosyl-transferase 2 protein-O-mannosyltransferase 2; Dolichyl-phosphate-mannose--protein mannosyltransferase; Dolichyl-phosphate-mannose--protein mannosyltransferase 2; Protein O-mannosyl-transferase 2; protein-O-mannosyltransferase 2
Gene Aliases: LGMD2N; MDDGA2; MDDGB2; MDDGC2; POMT2
UniProt ID: (Human) Q9UKY4
Entrez Gene ID: (Human) 29954
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.