Invitrogen
Manufacturer Code:PA542684
Catalog # PIPA542684
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human POMT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dolichyl-phosphate-mannose--protein mannosyltransferase 1; Protein O-mannosyl-transferase 1; protein O-mannosyltransferase 1; protein-O-mannosyltransferase 1; Protein-O-mannosyltransferase 1 FLJ37239 LGMD2K RT MDDGA1 MDDGB1 MDDGC1; testis tissue sperm-binding protein Li 57p
Gene Aliases: AI505244; LGMD2K; MDDGA1; MDDGB1; MDDGC1; POMT1; RT
UniProt ID: (Human) Q9Y6A1, (Rat) Q99PR0, (Mouse) Q8R2R1
Entrez Gene ID: (Horse) 100069575, (Dog) 608039, (Rabbit) 100357781, (Pig) 100525803, (Human) 10585, (Bovine) 617609, (Rat) 84430, (Mouse) 99011
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.