Novus Biologicals
Manufacturer Code:NBP29341320UL
Catalog # NB028709
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 310-550 of human POMT1 (NP_001070833.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dolichyl-phosphate-mannose--protein mannosyltransferase 1; Dolichyl-phosphate-mannose--protein mannosyltransferase 1 EC 2.4.1 EC 2.4.1.109 FLJ37239 LGMD2K MDDGA1 MDDGB1 MDDGC1 protein O-mannosyl-transferase 1 protein-O-mannosyltransferase 1 RT; Protein O-mannosyl-transferase 1; protein-O-mannosyltransferase 1; testis tissue sperm-binding protein Li 57p
Gene Aliases: LGMD2K; MDDGA1; MDDGB1; MDDGC1; POMT1; RT
UniProt ID: (Human) Q9Y6A1
Entrez Gene ID: (Human) 10585
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.