Invitrogen
Manufacturer Code:PA543488
Catalog # PIPA543488
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human FLJ23356 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: POMK; probable inactive protein kinase-like protein SgK196; Protein kinase-like protein SgK196; protein kinase-like protein SgK196-like; Protein O-mannose kinase; Sugen kinase 196
Gene Aliases: 4930444A02Rik; MDDGA12; MDDGC12; POMK; RGD1310810; SGK196
UniProt ID: (Human) Q9H5K3, (Rat) Q4V8A9, (Mouse) Q3TUA9
Entrez Gene ID: (Human) 84197, (Rabbit) 100346922, (Bovine) 514490, (Dog) 482834, (Rat) 306549, (Mouse) 74653, (Guinea pig) 100718330
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.