Invitrogen
Manufacturer Code:PA560735
Catalog # PIPA560735
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human POMGNT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GnT I.2; POMGnT1; Protein O-linked-mannose beta-1,2-N-acetylglucosaminyltransferase 1; RP11-322N21.3 GNTI.2 GnT I.2 MDDGA3 MDDGB3 MDDGC3 MEB MGAT1.2 gnT-I.2 UDP-GlcNAc:alpha-D-mannoside beta-12-N-acetylglucosaminyltransferase I.2 protein O-linked-mannose beta-12-N-acetylglucosaminyltransferase 1; UDP-GlcNAc:alpha-D-mannoside beta-1,2-N-acetylglucosaminyltransferase I.2
Gene Aliases: GnT I.2; gnT-I.2; GNTI.2; LGMD2O; MEB; MGAT1.2; POMGNT1; UNQ746/PRO1475
UniProt ID: (Human) Q8WZA1
Entrez Gene ID: (Human) 55624
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.