Invitrogen
Manufacturer Code:PA5117164
Catalog # PIPA5117164
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 411-660 of human POMGNT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610016I07Rik 4930467B06Rik GnT I.2 GNTI.2 gnT-I.2 LGMD2O MDDGA3 MDDGB3 MDDGC3 MEB MGAT1.2 O-linked mannose beta12-N-acetylglucosaminyltransferase O-mannosyl N-acetylglucosaminyltransferase OMGnT1 Pomgnt1 protein O-linked mannose beta 12-N-acetylglucosaminyltransferase protein O-linked mannose beta12-N-acetylglucosaminyltransferase protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 12-) protein O-linked-mannose beta-12-N-acetylglucosaminyltransferase 1 RP11-322N21.3 UDP-GlcNAc:alpha-D-mannoside beta-12-N-acetylglucosaminyltransferase I.2 UNQ746/PRO1475; GnT I.2; O-linked mannose beta1,2-N-acetylglucosaminyltransferase; O-mannosyl N-acetylglucosaminyltransferase; POMGnT1; protein O-linked mannose beta1,2-N-acetylglucosaminyltransferase; Protein O-linked-mannose beta-1,2-N-acetylglucosaminyltransferase 1; UDP-GlcNAc:alpha-D-mannoside beta-1,2-N-acetylglucosaminyltransferase I.2
Gene Aliases: 0610016I07Rik; 4930467B06Rik; GnT I.2; gnT-I.2; GNTI.2; LGMD2O; MEB; MGAT1.2; POMGNT1; UNQ746/PRO1475
UniProt ID: (Human) Q8WZA1, (Rat) Q5XIN7, (Mouse) Q91X88
Entrez Gene ID: (Human) 55624, (Rat) 362567, (Mouse) 68273
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.