Abnova
Manufacturer Code:MAB0905
Catalog # 89-108-233
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | B31.15 |
Immunogen | Native purified human POMC. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against native POMC.
For Research Use Only
Protein Aliases: ACTH; Adrenocorticotropic hormone; adrenocorticotropin; alpha-melanocyte-stimulating hormone; Alpha-MSH; Beta-endorphin; Beta-LPH; beta-melanocyte-stimulating hormone; Beta-MSH; CLIP; Corticotropin; Corticotropin-like intermediary peptide; Corticotropin-lipotropin; Gamma-LPH; Gamma-MSH; Lipotropin beta; Lipotropin gamma; LPH; Melanocyte-stimulating hormone alpha; Melanocyte-stimulating hormone beta; Melanotropin alpha; Melanotropin beta; Melanotropin gamma; Met-enkephalin; MSH; NPP; opiomelanocortin prepropeptide; POC; POMC; Potential peptide; pro-ACTH-endorphin; Pro-opiomelanocortin; proopiomelanocortin preproprotein
Gene Aliases: ACTH; CLIP; LPH; MSH; NPP; POC; POMC
UniProt ID: (Human) P01189
Entrez Gene ID: (Human) 5443
Molecular Function:
neuropeptide
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.