R&D Systems
Manufacturer Code:MAB9498SP
Catalog # MAB9498SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 967905 |
Immunogen | Human Adrenocorticotropic hormone/ACTH) synthetic peptide Accession # P01189 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: ACTH; ACTH adrenocorticotropic hormone adrenocorticotropin alpha-melanocyte-stimulating hormone alpha-MSH beta-endorphin beta-LPH beta-melanocyte-stimulating hormone beta-MSH CLIP corticotropin-like intermediary peptide corticotropin-lipotropin gamma-LPH gamma-MSH lipotropin beta lipotropin gamma LPH melanotropin alpha melanotropin beta melanotropin gamma met-enkephalin MSH NPP POC pro-ACTH-endorphin proopiomelanocortin pro-opiomelanocortin proopiomelanocortin preproprotein; Adrenocorticotropic hormone; adrenocorticotropin; alpha-melanocyte-stimulating hormone; Alpha-MSH; Beta-endorphin; Beta-LPH; beta-melanocyte-stimulating hormone; Beta-MSH; CLIP; Corticotropin; Corticotropin-like intermediary peptide; Corticotropin-lipotropin; Gamma-LPH; Gamma-MSH; Lipotropin beta; Lipotropin gamma; Melanocyte-stimulating hormone alpha; Melanocyte-stimulating hormone beta; Melanotropin alpha; Melanotropin beta; Melanotropin gamma; Met-enkephalin; NPP; opiomelanocortin prepropeptide; POMC; Potential peptide; pro-ACTH-endorphin; Pro-opiomelanocortin; proopiomelanocortin preproprotein
Gene Aliases: ACTH; CLIP; LPH; MSH; NPP; POC; POMC
UniProt ID: (Human) P01189
Entrez Gene ID: (Human) 5443
Molecular Function:
neuropeptide
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.