Invitrogen
Manufacturer Code:PA185109
Catalog # PIPA185109
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the amino acids 1-24 of ACTH conjugated to Keyhole limpet Haemocyanin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4°C or -20°C if preferred |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACTH; Adrenocorticotropic hormone; adrenocorticotropin; alpha-melanocyte-stimulating hormone; Alpha-MSH; Beta-endorphin; Beta-LPH; beta-melanocyte-stimulating hormone; Beta-MSH; CLIP; Corticotropin; Corticotropin-like intermediary peptide; Corticotropin-lipotropin; Gamma-LPH; Gamma-MSH; Lipotropin beta; Lipotropin gamma; Melanocyte-stimulating hormone alpha; Melanocyte-stimulating hormone beta; Melanotropin alpha; Melanotropin beta; Melanotropin gamma; Met-enkephalin; NPP; opiomelanocortin prepropeptide; POMC; Potential peptide; pro-ACTH-endorphin; Pro-opiomelanocortin; pro-opiomelanocortin-alpha; proopiomelanocortin (adrenocorticotropin/ beta-lipotropin/ alpha-melanocyte stimulating hormone/ beta-melanocyte stimulating hormone/ beta-endorphin); proopiomelanocortin preproprotein; proopiomelanocortin, beta (endorphin, beta); proopoimelanocortin, beta (endorphin, beta)
Gene Aliases: ACTH; alphaMSH; CLIP; LPH; MSH; NPP; POC; POMC; Pomc1; Pomc2
UniProt ID: (Human) P01189
Entrez Gene ID: (Human) 5443, (Rat) 24664
Molecular Function: neuropeptide peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.