Invitrogen
Manufacturer Code:PA585161
Catalog # PIPA585161
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the N-terminus region of human POM121. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp586G1822 antibody DKFZp586P2220 antibody FLJ41820 antibody KIAA0618 antibody MGC3792 antibody POM121A antibody POM121 antibody P145 antibody pore membrane protein of 121 kDa antibody nuclear envelope pore membrane protein POM 121 antibody nuclear envelope pore membrane protein POM 121A antibody nucleoporin Nup121 antibody nuclear pore membrane protein 121 kDa antibody POM121 membrane glycoprotein antibody; Nuclear envelope pore membrane protein POM 121; Nuclear envelope pore membrane protein POM 121A; nuclear pore membrane protein 121 kDa; Nucleoporin Nup121; POM121 membrane glycoprotein; Pore membrane protein of 121 kDa
Gene Aliases: 2610027A18Rik; C80273; KIAA0618; mKIAA0618; NUP121; P145; POM121; POM121A
UniProt ID: (Human) Q96HA1, (Mouse) Q8K3Z9
Entrez Gene ID: (Human) 9883, (Mouse) 107939
Molecular Function: RNA binding protein nucleic acid binding structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.