OriGene
Manufacturer Code:TA809432
Catalog # 50-168-2786
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4D8 |
Immunogen | Full length human recombit protein of human POLR3H produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DNA-directed RNA polymerase III subunit 22.9 kDa polypeptide; DNA-directed RNA polymerase III subunit H; DNA-directed RNA polymerase III subunit RPC8; polymerase (RNA) III (DNA directed) polypeptide H (22.9kD); polymerase (RNA) III subunit H; RNA nucleotidyltransferase (DNA-directed); RNA polymerase III subunit 22.9 kDa subunit; RNA polymerase III subunit C8; RPC22.9; RPC22.9 RPC8
Gene Aliases: KIAA1665; POLR3H; RPC22.9; RPC8
UniProt ID: (Human) Q9Y535
Entrez Gene ID: (Human) 171568
Molecular Function: DNA-directed RNA polymerase RNA binding protein nucleic acid binding nucleotidyltransferase transcription factor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.