Invitrogen
Manufacturer Code:PA555640
Catalog # PIPA555640
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human POLR3C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA-directed III 62 kDa polypeptide; DNA-directed RNA polymerase III subunit C; DNA-directed RNA polymerase III subunit RPC3; polymerase (RNA) III (DNA directed) polypeptide C (62kD); Polymerase (RNA) III (DNA directed) polypeptide C (62kD) RPC3 RPC62; polymerase (RNA) III subunit C; RNA polymerase III 62 kDa subunit; RNA polymerase III subunit C3; RPC62
Gene Aliases: 4933407E01Rik; POLR3C; RPC3; RPC62
UniProt ID: (Human) Q9BUI4, (Mouse) Q9D483, (Rat) Q5XIL3
Entrez Gene ID: (Human) 10623, (Mouse) 74414, (Rat) 310685
Molecular Function:
DNA-directed RNA polymerase
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.