Novus Biologicals
Manufacturer Code:NBP154363
Catalog # NBP154363
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to POLR3B(polymerase (RNA) III (DNA directed) polypeptide B) The peptide sequence was selected from the C terminal of POLR3B. Peptide sequence IEKVMISSNAEDAFLIKMLLRQTRRPEIGDKFSSRHGQKGVCGLIVPQED. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C128; C128 DKFZp686B10117 DNA-directed RNA polymerase III 127.6 kDa polypeptide DNA-directed RNA polymerase III subunit B DNA-directed RNA polymerase III subunit RPC2 EC 2.7.7.6 FLJ10388 polymerase (RNA) III (DNA directed) polypeptide B RNA polymerase III subunit C2 RPC2; DNA-directed RNA polymerase III 127.6 kDa polypeptide; DNA-directed RNA polymerase III subunit B; DNA-directed RNA polymerase III subunit RPC2; interphase nucleus and mitotic apparatus associated protein; polymerase (RNA) III (DNA directed) polypeptide B; polymerase (RNA) III subunit B; RNA polymerase III subunit C2
Gene Aliases: C128; HLD8; INMAP; POLR3B; RPC2
UniProt ID: (Human) Q9NW08
Entrez Gene ID: (Human) 55703
Molecular Function: DNA-directed RNA polymerase RNA binding protein nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.