Invitrogen
Manufacturer Code:PA543213
Catalog # PIPA543213
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human POLR3A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA-directed RNA polymerase III largest subunit; DNA-directed RNA polymerase III subunit A; DNA-directed RNA polymerase III subunit RPC1; Polymerase (RNA) III (DNA directed) polypeptide A RPC1 RPC155 hRPC155 ADDH HLD7; polymerase (RNA) III (DNA directed) polypeptide A, 155kDa; polymerase (RNA) III subunit A; RNA polymerase III 155 kDa subunit; RNA polymerase III subunit C1; RNA polymerase III subunit C160; RNA polymerase III subunit RPC155-D; RPC155
Gene Aliases: 9330175N20Rik; ADDH; BC053071; HLD7; hRPC155; POLR3A; RGD1305574; RPC1; RPC155
UniProt ID: (Bovine) A4IF62, (Human) O14802
Entrez Gene ID: (Dog) 479256, (Rabbit) 100355224, (Horse) 100073003, (Bovine) 540308, (Human) 11128, (Mouse) 218832, (Rat) 361102, (Guinea pig) 100724395
Molecular Function:
DNA-directed RNA polymerase
RNA binding protein
nucleic acid binding
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.